Merck
HPLC Analysis of 2,2′-Bis[(S)-(4-methylphenyl)sulfinyl]-1,1′-binapthaIene Diastereomers on Ascentis® Express C18
HPLC Analysis of 2,2′-Bis[(S)-(4-methylphenyl)sulfinyl]-1,1′-binapthaIene Diastereomers on Ascentis® Express C18 application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Ascentis® Express C18, 2.7 μm HPLC Column
2.7 μm particle size, L × I.D. 10 cm × 4.6 mm
standard
Product No.
Description
Pricing
(1S)-2,2′-Bis[(S)-(4-methylphenyl)sulfinyl]-1,1′-binaphthalene
≥96.5% (HPLC)
(1R)-2,2′-Bis[(S)-(4-methylphenyl)sulfinyl]-1,1′-binaphthalene
97%
CONDITIONS
column
Ascentis Express C18, 10 cm x 4.6 mm I.D., 2.7 μm particles (53827-U)
mobile phase
[A] water; [B] methanol; (25:75, A:B)
flow rate
0.8 mL/min
pressure
2450 psi (169 bar)
column temp.
35 °C
detector
UV, 240 nm
injection
5 μL
sample
(1R)-2,2′-Bis[(S )-(4-methylphenyl)sulfonyl]-1,1′-binapthalene, 25 μg/mL in methanol; (1S)-2,2′-Bis[(S)-(4-methylphenyl)sulfonyl]-1,1′-binapthalene, 50 μg/mL in methanol
Description
Analysis Note
This application demonstrates the suitability of the Ascentis Express C18 for the separation of the diastereomers of by HPLC in reversed phase mode.
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany