Merck
Base-Resolution Mapping Reveals Distinct m
Molecular cell (2017-11-07)
Xiaoyu Li, Xushen Xiong, Meiling Zhang, Kun Wang, Ying Chen, Jun Zhou, Yuanhui Mao, Jia Lv, Danyang Yi, Xiao-Wei Chen, Chu Wang, Shu-Bing Qian, Chengqi Yi
ABSTRACT
Gene expression can be post-transcriptionally regulated via dynamic and reversible RNA modifications. N
Sigma-Aldrich
MISSION® esiRNA, targeting human TRMT61B
Sigma-Aldrich
Nuclease P1 from Penicillium citrinum, lyophilized powder, ≥200 units/mg protein (E1%/280, 3′-5′-Phosphodiesterase)
Sigma-Aldrich
Cy3-alkyne, for copper catalyzed click labeling
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, ammonium sulfate suspension, 30-90 units/mg protein (modified Warburg-Christian, in glycine buffer)
Sigma-Aldrich
Emetine dihydrochloride