Merck
  • Efficient and specific gene knockdown by small interfering RNAs produced in bacteria.
Efficient and specific gene knockdown by small interfering RNAs produced in bacteria.
Nature biotechnology (2013-03-12)
Linfeng Huang, Jingmin Jin, Padraig Deighan, Evgeny Kiner, Larry McReynolds, Judy Lieberman
ABSTRACT
Synthetic small interfering RNAs (siRNAs) are an indispensable tool to investigate gene function in eukaryotic cells and may be used for therapeutic purposes to knock down genes implicated in disease. Thus far, most synthetic siRNAs have been produced by chemical synthesis. Here we present a method to produce highly potent siRNAs in Escherichia coli. This method relies on ectopic expression of p19, an siRNA-binding protein found in a plant RNA virus. When expressed in E. coli, p19 stabilizes an ∼21-nt siRNA-like species produced by bacterial RNase III. When mammalian cells are transfected by them, siRNAs that were generated in bacteria expressing p19 and a hairpin RNA encoding 200 or more nucleotides of a target gene reproducibly knock down target gene expression by ∼90% without immunogenicity or off-target effects. Because bacterially produced siRNAs contain multiple sequences against a target gene, they may be especially useful for suppressing polymorphic cellular or viral genes.
Sigma-Aldrich
Anti-pan Ago Antibody, clone 2A8, clone 2A8, from mouse
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)