Merck
  • Basal Autophagy and Feedback Activation of Akt Are Associated with Resistance to Metformin-Induced Inhibition of Hepatic Tumor Cell Growth.
Basal Autophagy and Feedback Activation of Akt Are Associated with Resistance to Metformin-Induced Inhibition of Hepatic Tumor Cell Growth.
PloS one (2015-06-26)
Hua Yang, Yuan-Fei Peng, Hong-Min Ni, Yuan Li, Ying-Hong Shi, Wen-Xing Ding, Jia Fan
ABSTRACT
While accumulating evidence has shown that the use of the diabetic drug metformin may be beneficial against various tumors in some epidemiological studies, a few studies failed to show the same beneficial effects. The molecular and cellular mechanisms for these conflicting observations are not clear. In this study, we compared the inhibitory effects of cell growth by metformin on several hepatic tumor cell lines: SMMC-7721, HCC-97L, HCC-LM3 and HepG2. While metformin inhibited cell growth in all these cells, we found that SMMC-7721, HCC-97L and HCC-LM3 cells were more resistant than HepG2 cells. Mechanistically, we found that metformin inhibited mTOR in all these hepatic tumor cells. However, SMMC-7721 cells had higher levels of basal autophagy and mTORC2-mediated feedback activation of Akt than HepG2 cells, which may render SMMC-7721 cells to be more resistant to metformin-induced inhibition of cell growth. Similarly, HCC-97L and HCC-LM3 cells also had higher feedback activation of AKT than HepG2 cells, which may also account for their resistance to metformin-induced inhibition of cell growth. Therefore, the various basal autophagy and mTOR activity in different cancer cells may contribute to the controversial findings on the use of metformin in inhibition of cancers in humans.
Sigma-Aldrich
Rapamycin, Ready Made Solution, 2.5 mg/mL in DMSO (2.74 mM), from Streptomyces hygroscopicus
Sigma-Aldrich
N-Acetyl-Asp-Glu-Val-Asp-7-amido-4-trifluoromethylcoumarin, ≥90% (HPLC), powder
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid