Merck
Methods for recovering nucleic acid fragments from agarose gels.
Journal of chromatography (1993-08-25)
G Duro, V Izzo, R Barbieri
ABSTRACT
Agarose gel electrophoresis is a powerful technique for the separation of nucleic acids on the basis of their size and conformation. The development of methods to recover size-fractionated nucleic acids molecules from agarose gels has greatly facilitated recombinant DNA technologies. Although several methods for recovering DNA and RNA molecules have been developed during the past fifteen years, none of them has been universally accepted. In this review we describe, discuss and evaluate the most common procedures with which we have had experience. Our evaluation is based on the criteria of yield, purity, speed, simplicity and low cost. We have considered three different approaches to the problem of recovering nucleic acids: chemical gel dissolution, physical gel disruption and physical extrusion from intact gels.
Sigma-Aldrich
Agarose, High EEO
Sigma-Aldrich
Agarose, Ultra-low Gelling Temperature, molecular biology grade
Sigma-Aldrich
Agarose, Type II-A, Medium EEO
Sigma-Aldrich
Agarose, Low EEO
Sigma-Aldrich
Agarose, for molecular biology
Sigma-Aldrich
Agarose, Ultra-low Gelling Temperature
Sigma-Aldrich
Sepharose 6B, 6% Beaded Agarose, 45-165 μm (wet), fractionation range 10,000-1,000,000 Da (dextrans)
Sigma-Aldrich
Agarose, High Gelling Temperature
Sigma-Aldrich
Agarose, Type I, low EEO
Sigma-Aldrich
Agarose, Wide range, for molecular biology
Sigma-Aldrich
Agarose, For pulsed field electrophoresis running gel
Sigma-Aldrich
Agarose, BioReagent, for molecular biology, low EEO
Sigma-Aldrich
Agarose, Medium EEO, for molecular biology
Sigma-Aldrich
Agarose, Type I-A, low EEO
Sigma-Aldrich
Agarose, Type IV, Special High EEO
Sigma-Aldrich
Agarose, BioReagent, for molecular biology, Wide range/Standard 3:1
Sigma-Aldrich
Agarose, Low EEO, for Immunoelectrophoresis
Sigma-Aldrich
Agarose, High EEO, for molecular biology
Sigma-Aldrich
Agarose, Special High EEO
Sepharose 6B, Cytiva 17-0110-01, pack of 1 L
Sigma-Aldrich
Sepharose 4B, 45-165 μm bead diameter
Sepharose 4B, Cytiva 17-0120-01, pack of 1 L