Merck
  • New developments in medulloblastoma treatment: the potential of a cyclopamine-lovastatin combination.
New developments in medulloblastoma treatment: the potential of a cyclopamine-lovastatin combination.
Expert opinion on investigational drugs (2008-01-31)
Eli E Bar, Duncan Stearns
ABSTRACT
Pediatric brain tumors are the most common cause of cancer-related death in children. Despite advances in neuroimaging, surgical techniques, radiotherapy and the introduction of new chemotherapeutic agents, up to 30% of all patients diagnosed with medulloblastoma will die from their disease, thus underscoring the need for more effective treatments. This review explores the relationship between cholesterol and cancer in light of a recent study that demonstrated that the hedgehog (Hh) antagonist cyclopamine blocked expression of the Hh pathway targets PTCH1 and GLI1, lowered Bcl2 levels and increased apoptosis in medulloblastoma cells. In addition, Hh pathway blockade sensitized medulloblastoma to the effects of the proapoptotic, cholesterol-lowering agent lovastatin. The use of Hh pathway antagonists and cholesterol synthesis inhibitors in medulloblastoma is a novel and exciting approach as it targets pathways crucial for tumor survival. The safety of such approach, however, still needs to be tested in animal models of the disease before its application in human patients.
Product Description
Lovastatin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Mevinolin from Aspergillus sp., ≥98% (HPLC)
Supelco
Lovastatin, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Lovastatin, United States Pharmacopeia (USP) Reference Standard