Merck
[Use of cefuroxime axetil in clinical practice].
Polski merkuriusz lekarski : organ Polskiego Towarzystwa Lekarskiego (2001-02-24)
M Słowińska, E Szymańska, I Walecka, L Rudnicka
ABSTRACT
This study summarizes the actual recommendations for cefuroxime axetil treatment in dermatology and general medicine. These include the well known clinical efficacies in therapy of upper and lower respiratory tract infections, genitourinary tract infections and skin and soft tissue infections, but also connective tissue diseases, such as morphea and SCLE. Though the immunomodulatory activity of the drug should be established by further controlled studies, there are some limited literature data, which show the modulatory effect of cefuroxime axetil on the lymphocyte proliferation and the production of selected cytokines.
Product Description
Cefuroxime Axetil, European Pharmacopoeia (EP) Reference Standard
USP
Cefuroxime Axetil, United States Pharmacopeia (USP) Reference Standard
Supelco
Cefuroxime, VETRANAL®, analytical standard