Merck
  • Methotrexate and lung disease in rheumatoid arthritis: a meta-analysis of randomized controlled trials.
Methotrexate and lung disease in rheumatoid arthritis: a meta-analysis of randomized controlled trials.
Arthritis & rheumatology (Hoboken, N.J.) (2014-04-24)
Richard Conway, Candice Low, Robert J Coughlan, Martin J O'Donnell, John J Carey
ABSTRACT
Methotrexate has shown efficacy for the treatment of several diseases, especially rheumatoid arthritis (RA). Methotrexate has also been implicated as a causative agent in interstitial lung disease. Patients with RA may develop pulmonary manifestations of their disease and are at increased risk of respiratory infection. The aim of this study was to evaluate the relative risk (RR) of pulmonary disease among patients with RA treated with methotrexate. We searched the PubMed and Cochrane databases (publication dates January 1, 1990 to February 1, 2013) for double-blind, randomized, controlled trials of methotrexate versus placebo or active comparator agents in adults with RA. Studies with <100 subjects or with a duration of <24 weeks were excluded. Two investigators independently searched both databases, and all of the investigators reviewed the selected studies. We compared differences in the RR using the Mantel-Haenszel random-effects method. A total of 22 studies with 8,584 participants met the inclusion criteria. Heterogeneity across studies was not significant (I(2) = 3%), allowing combination of the trial results. Methotrexate was associated with an increased risk of all adverse respiratory events (RR 1.10, 95% confidence interval [95% CI] 1.02-1.19) and respiratory infection (RR 1.11, 95% CI 1.02-1.21). Patients treated with methotrexate were not at increased risk of death due to lung disease (RR 1.53, 95% CI 0.46-5.01) or noninfectious respiratory events (RR 1.02, 95% CI 0.65-1.60). A subgroup analysis of studies in which pneumonitis was described revealed an increased risk associated with methotrexate (RR 7.81, 95% CI 1.76-34.72). Our study demonstrated a small but significant increase in the risk of lung disease in patients with RA treated with methotrexate compared with other disease-modifying antirheumatic drugs and biologic agents.
Product Description
Methotrexate for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Methotrexate hydrate, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
Methotrexate hydrate, ≥98% (HPLC), powder
Methotrexate for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Methotrexate hydrate, powder, BioReagent, suitable for cell culture, ≥98% (HPLC)
Sigma-Aldrich
Methotrexate, meets USP testing specifications
Supelco
Methotrexate solution, 1.0 mg/mL in methanol with 0.1N NaOH, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Methotrexate, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Methotrexate
Methotrexate, European Pharmacopoeia (EP) Reference Standard