Sigma-Aldrich
Hydroxyapatite, purum p.a., ≥90% (as Ca3(PO4)2, KT)
Sigma-Aldrich
Hydroxyapatite, puriss., meets analytical specification of Ph. Eur., BP, FCC, E341, ≥90% (calculated on glowed substance)
Sigma-Aldrich
Silicon, wafer, <111>, P-type, contains boron as dopant, diam. × thickness 2 in. × 0.3 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <111>, N-type, contains no dopant, diam. × thickness 2 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <111>, N-type, contains no dopant, diam. × thickness 3 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <100>, N-type, contains no dopant, diam. × thickness 3 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <100>, N-type, contains phosphorus as dopant, diam. × thickness 2 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <111>, N-type, contains phosphorus as dopant, diam. × thickness 2 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <111>, P-type, contains boron as dopant, diam. × thickness 3 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <100>, N-type, contains no dopant, diam. × thickness 2 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <100>, P-type, contains boron as dopant, diam. × thickness 2 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <100>, N-type, contains phosphorus as dopant, diam. × thickness 3 in. × 0.5 mm
Sigma-Aldrich
Silicon, nanopowder, <100 nm particle size (TEM), ≥98% trace metals basis
Sigma-Aldrich
Silicon, pieces, 99.95% trace metals basis
Sigma-Aldrich
Silicon, powder, −325 mesh, 99% trace metals basis
Sigma-Aldrich
Silicon, wafer (single side polished), <100>, P-type, contains boron as dopant, diam. × thickness 3 in. × 0.5 mm
Silicon, rod, 40mm, diameter 20mm, single crystal, -100, 99.999%
Silicon, sheet, 10x10mm, thickness 0.5mm, single crystal, -100, 100%
Silicon, rod, 50mm, diameter 2.0mm, crystalline, 100%
Silicon, sheet, 14x14mm, thickness 1.0mm, single crystal, -111, 100%
Silicon, sheet, 10x10mm, thickness 0.6mm, single crystal, -100, 100%
Silicon, rod, 50mm, diameter 3.15mm, single crystal - random orientation, 100%
Silicon, rod, 50mm, diameter 5.0mm, crystalline, 100%
Silicon, rod, 50mm, diameter 5mm, single crystal, -100, 99.999%
Silicon, rod, 100mm, diameter 5.0mm, crystalline, 100%
Silicon, rod, 25mm, diameter 3.15mm, single crystal - random orientation, 100%
Silicon, sheet, 25x25mm, thickness 1.0mm, single crystal, -100, 100%
Silicon, sheet, 52x52mm, thickness 1.0mm, polycrystalline, 99.999%
Silicon, rod, 10mm, diameter 2.0mm, crystalline, 100%
Silicon, disks, 15.9mm, thickness 0.38mm, single crystal, n-type, 100%