Merck
  • Network state-dependent inhibition of identified hippocampal CA3 axo-axonic cells in vivo.
Network state-dependent inhibition of identified hippocampal CA3 axo-axonic cells in vivo.
Nature neuroscience (2013-10-22)
Tim J Viney, Balint Lasztoczi, Linda Katona, Michael G Crump, John J Tukker, Thomas Klausberger, Peter Somogyi
ABSTRACT
Hippocampal sharp waves are population discharges initiated by an unknown mechanism in pyramidal cell networks of CA3. Axo-axonic cells (AACs) regulate action potential generation through GABAergic synapses on the axon initial segment. We found that CA3 AACs in anesthetized rats and AACs in freely moving rats stopped firing during sharp waves, when pyramidal cells fire most. AACs fired strongly and rhythmically around the peak of theta oscillations, when pyramidal cells fire at low probability. Distinguishing AACs from other parvalbumin-expressing interneurons by their lack of detectable SATB1 transcription factor immunoreactivity, we discovered a somatic GABAergic input originating from the medial septum that preferentially targets AACs. We recorded septo-hippocampal GABAergic cells that were activated during hippocampal sharp waves and projected to CA3. We hypothesize that inhibition of AACs, and the resulting subcellular redistribution of inhibition from the axon initial segment to other pyramidal cell domains, is a necessary condition for the emergence of sharp waves promoting memory consolidation.
Sigma-Aldrich
Biotin, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Biotin, meets USP testing specifications
Sigma-Aldrich
Biotin, tested according to Ph. Eur.
Biotin, European Pharmacopoeia (EP) Reference Standard
Supelco
Biotin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Biotin, ≥99.0% (T)
Supelco
Biotin, Pharmaceutical Secondary Standard; Certified Reference Material