Merck
  • Benzophenone-type UV filters in urine and blood from children, adults, and pregnant women in China: partitioning between blood and urine as well as maternal and fetal cord blood.
Benzophenone-type UV filters in urine and blood from children, adults, and pregnant women in China: partitioning between blood and urine as well as maternal and fetal cord blood.
The Science of the total environment (2013-05-29)
Tao Zhang, Hongwen Sun, Xiaolei Qin, Qian Wu, Yanfeng Zhang, Jing Ma, Kurunthachalam Kannan
ABSTRACT
Limited information exists on the exposure of benzophenone (BP)-type UV filters (i.e., sunscreen compounds) in children, adults, and pregnant women in China. In this study, we determined the concentrations of five BP derivatives, BP-1, BP-2, BP-3, BP-8, and 4OH-BP in urine (n=101) as well as paired specimens of blood and urine (n=24 pairs) collected from adults; in matched maternal and fetal cord blood (n=20 pairs) collected from pregnant women; and in blood collected from children (n=10). 4OH-BP, BP-1, and BP-3 were found in 61%, 57%, and 25%, respectively, of the urine samples analyzed. 4OH-BP was found in all blood samples; BP-3 was found more frequently in the blood of adults (83%), followed, in decreasing order, by pregnant women (35%) and children (30%). Among all adults, urinary BP-3 concentrations were significantly (p<0.001) positively correlated with urinary BP-1 concentrations. Nevertheless, no significant correlations were found between urinary concentrations of BP-3 (or BP-1) and 4OH-BP. Our results suggest that human exposure to BP-3 and BP-1 is related, whereas 4OH-BP originates from a discrete source. Females had higher urinary concentrations of BP-3, BP-1 and 4OH-BP than males. The distribution profiles of BP-1 and its parent compound (i.e., BP-3) in urine decreased with increasing age of donors (p<0.05). The ratio of concentrations of BP-3 between blood and urine was 0.21 in adults, which was significantly lower than that for 4OH-BP (0.36). The concentration ratio of BPs between cord blood and maternal blood was higher for 4OH-BP (0.61) than that for BP-3 (0.48), which suggested greater trans-placental transfer potential of 4OH-BP. This is the first study to document the occurrence of BPs in paired urine and blood, and in matched maternal and fetal cord blood.
Product Description
Supelco
Melting point standard 47-49°C, analytical standard
Supelco
Mettler-Toledo Calibration substance ME 18870, Benzophenone, traceable to primary standards (LGC)
Supelco
Benzophenone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Benzophenone, purified by sublimation, ≥99%
Supelco
Benzophenone, analytical standard
Sigma-Aldrich
Benzophenone, ReagentPlus®, 99%
Benzophenone, European Pharmacopoeia (EP) Reference Standard