Merck
  • Responsiveness to three bronchial provocation tests in patients with asthma.
Responsiveness to three bronchial provocation tests in patients with asthma.
Chest (2003-12-11)
Heikki O Koskela, Liisa Hyvärinen, John D Brannan, Hak-Kim Chan, Sandra D Anderson
ABSTRACT
To compare a new bronchial provocation test, the mannitol challenge, with cold air and histamine challenges to demonstrate airway hyperresponsiveness (AHR) in patients with difficult-to-diagnose asthma. A prospective study. Thirty-seven consecutive patients with recently diagnosed, steroid-naive, mild, or atypical asthma fulfilling the diagnostic criteria of Finnish Social Insurance Institution, and 10 healthy control subjects. Each subject completed a symptom questionnaire and underwent spirometry, diffusion capacity measurement, skin-prick tests, and bronchial provocations with mannitol, histamine, and cold air. The severity of asthma was classified according to the Global Initiative for Asthma (GINA). Fifty-one percent of the asthmatic patients responded to mannitol (> or =15% fall in FEV(1)), 24% to cold air (> or =9% fall in FEV(1)), and 81% or 49% to histamine (provocative dose causing a 15% fall in FEV(1) [PD(15)] < 1.0 mg or < 0.4 mg, respectively). None of the healthy control subjects responded. The GINA classification was not associated with responsiveness to any of the challenges. Mannitol is more sensitive than cold air in demonstrating AHR in patients with mild or atypical asthma. Histamine was more sensitive than both mannitol and cold air if 1.0 mg was used as a cut-off value for histamine PD(15). However, if the cut-off value for histamine PD(15) is lowered to 0.4 mg, which represents a specific diagnosis of asthma according to previous studies, the sensitivity values of mannitol and histamine challenges are comparable.
Product Description
Mannitol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-Mannitol, tested according to Ph. Eur.
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Millipore
D-Mannitol, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
Supelco
Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material