Merck
  • Application of XAD-2 resin-based passive samplers and SPME-GC-MS/MS analysis for the monitoring of spatial and temporal variations of atmospheric pesticides in Luxembourg.
Application of XAD-2 resin-based passive samplers and SPME-GC-MS/MS analysis for the monitoring of spatial and temporal variations of atmospheric pesticides in Luxembourg.
Environmental pollution (Barking, Essex : 1987) (2012-07-10)
Claude Schummer, Ludovic Tuduri, Olivier Briand, Brice M Appenzeller, Maurice Millet
ABSTRACT
Passive air sampling has been shown to be a very interesting alternative to high-volume sampling by overcoming its disadvantages (size, weight, expensiveness). However, to date, only limited data is available about passive air sampling of current-use pesticides. In order to test if passive samplers allow monitoring of spatial and temporal variations of atmospheric pesticide concentrations, five XAD-2-resin based passive air samplers were deployed at five locations in Luxembourg. Samplers were analyzed using accelerated solvent extraction coupled to solid-phase microextraction and gas chromatography with tandem mass spectrometry. Collected data was used to study the spatial and temporal variations of the concentrations of the compounds. Twenty two pesticides were detected between March and October, while no pesticides were detected from November to February. Highest concentrations were measured on the rural sites, suggesting that the used XAD-2 resin-based passive samplers allow the simultaneous monitoring of multiple current-use pesticides and identifying spatial and temporal variations.
Product Description
Supelco
Amberlite XAD®-2, pkg of 100 g
Supelco
Amberlite XAD®-2, pkg of 25 kg
Supelco
Amberlite XAD®-2, pkg of 10 kg
Supelco
Amberlite XAD®-2, pkg of 500 g
Supelco
Amberlite XAD®-2, pkg of 5 kg