- High temperature purge and trap procedure for determining seven volatile N-nitrosamines in animal feed, using gas chromatography/thermal energy analyzer.
High temperature purge and trap procedure for determining seven volatile N-nitrosamines in animal feed, using gas chromatography/thermal energy analyzer.
A rapid and sensitive procedure is described for determining 4 N-nitrosodialkylamines (dimethyl, diethyl, dipropyl, and dibutyl) and the N-nitroso analogs of piperidine, pyrrolidine, and morpholine in animal feed. The volatile N-nitrosamines were isolated by using a modified high temperature purge and trap apparatus designed for multiple sample cleanup. The feed sample was mixed with mineral oil and a nitrosation inhibitor, and the N-nitrosamines were purged onto a ThermoSorb/N cartridge, eluted with acetone-dichloromethane (1 + 1), and determined by gas chromatography, using a thermal energy analyzer. The effects of purge rate, purge time, and temperature on recovery are discussed. Feed samples spiked with 10, 50, and 200 ppb of the 7 N-nitroso compounds yielded recoveries ranging from 70 to 97%, with precision ranging from 1.3 to 5.4% and minimum detectable levels in the low parts-per-billion range.