Merck
  • [Comparative studies on the genotoxic activity of a new palladium (II) acidocomplex vs cisplatin in human blood lymphocytes in vitro].
[Comparative studies on the genotoxic activity of a new palladium (II) acidocomplex vs cisplatin in human blood lymphocytes in vitro].
Biomeditsinskaia khimiia (2013-05-09)
A K Grekhova, L B Gorbacheva, N A Ivanova, I A Efimenko, A N Osipov
ABSTRACT
A comparative study on the genotoxic activity of cisplatin versus morfozol, the first representative of a new class of cation-anion complexes of palladium [AH]2[PdCl4] (where A-methylmorpholine) has been performed using human lymphocytes in vitro. The results of the DNA-DNA cross-linking activity investigations showed that both compounds studied exhibited biphasic dose-effect relationship: a linear decrease in the DNA percent in the comet tail and the region of the "plateau". However, in the "plateau" region, morfozol reduced the DNA percent in the comet tail up to 6 times while cisplatin caused a 2-fold decrease only. Morfozol, like cisplatin, inducing DNA-protein cross-linking and generating reactive oxygen species, was more effective than cisplatin.
Sigma-Aldrich
Palladium(II) chloride solution, 5 wt. % in 10 wt. % HCl
Sigma-Aldrich
Palladium(II) chloride ChemBeads
Sigma-Aldrich
Palladium, powder, <75 μm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, powder or granules, 99.99% trace metals basis
Sigma-Aldrich
Palladium, foil, thickness 1.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, foil, thickness 0.25 mm, 99.98% trace metals basis
Sigma-Aldrich
Palladium, wire, diam. 0.5 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium(II) chloride, anhydrous, 59-60% palladium (Pd) basis
Sigma-Aldrich
Palladium(II) chloride, 99.995%
Sigma-Aldrich
Palladium, powder, <1 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Palladium, foil, thickness 0.025 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, wire, diam. 1.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium(II) chloride, ≥99.9%
Sigma-Aldrich
Palladium, evaporation slug, diam. × L 0.6 cm × 0.6 cm, 99.95% trace metals basis
Sigma-Aldrich
Palladium(II) chloride, ReagentPlus®, 99%
Sigma-Aldrich
Palladium, sponge, 99.9% trace metals basis
Sigma-Aldrich
Palladium(II) chloride, anhydrous, 60% Pd basis
Sigma-Aldrich
Palladium, powder, 99.995% trace metals basis
Sigma-Aldrich
Palladium, foil, thickness 0.5 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, nanopowder, <25 nm particle size (TEM), ≥99.5%