Merck
  • Temporal decoding of MAP kinase and CREB phosphorylation by selective immediate early gene expression.
Temporal decoding of MAP kinase and CREB phosphorylation by selective immediate early gene expression.
PloS one (2013-03-08)
Takeshi H Saito, Shinsuke Uda, Takaho Tsuchiya, Yu-ichi Ozaki, Shinya Kuroda
ABSTRACT
A wide range of growth factors encode information into specific temporal patterns of MAP kinase (MAPK) and CREB phosphorylation, which are further decoded by expression of immediate early gene products (IEGs) to exert biological functions. However, the IEG decoding system remain unknown. We built a data-driven based on time courses of MAPK and CREB phosphorylation and IEG expression in response to various growth factors to identify how signal is processed. We found that IEG expression uses common decoding systems regardless of growth factors and expression of each IEG differs in upstream dependency, switch-like response, and linear temporal filters. Pulsatile ERK phosphorylation was selectively decoded by expression of EGR1 rather than c-FOS. Conjunctive NGF and PACAP stimulation was selectively decoded by synergistic JUNB expression through switch-like response to c-FOS. Thus, specific temporal patterns and combinations of MAPKs and CREB phosphorylation can be decoded by selective IEG expression via distinct temporal filters and switch-like responses. The data-driven modeling is versatile for analysis of signal processing and does not require detailed prior knowledge of pathways.
Sigma-Aldrich
Nerve Growth Factor from Vipera lebetina venom, NGF, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Anisomycin, Ready Made Solution from Streptomyces griseolus, 10 mg/mL in DMSO
Sigma-Aldrich
Anisomycin from Streptomyces griseolus, ≥98% (HPLC), solid