Merck
  • Ethanol-based organosolv treatment with trace hydrochloric acid improves the enzymatic digestibility of Japanese cypress (Chamaecyparis obtusa) by exposing nanofibers on the surface.
Ethanol-based organosolv treatment with trace hydrochloric acid improves the enzymatic digestibility of Japanese cypress (Chamaecyparis obtusa) by exposing nanofibers on the surface.
Bioresource technology (2013-02-12)
Akihiro Hideno, Ayato Kawashima, Takashi Endo, Katsuhisa Honda, Masatoshi Morita
ABSTRACT
The effects of adding trace acids in ethanol based organosolv treatment were investigated to increase the enzymatic digestibility of Japanese cypress. A high glucose yield (60%) in the enzymatic hydrolysis was obtained by treating the sample at 170 °C for 45 min in 50% ethanol liquor containing 0.4% hydrochloric acid. Moreover, the enzymatic digestibility of the treated sample was improved to ∼70% by changing the enzyme from acremonium cellulase to Accellerase1500. Field emission scanning electron microscopy revealed the presence of lignin droplets and partial cellulose nanofibers on the surface of the treated sample. Simultaneous saccharification and fermentation of the treated samples using thermotolerant yeast (Kluyveromyces marxianus NBRC1777) was tested. A high ethanol concentration (22.1 g/L) was achieved using the EtOH50/W50/HCl0.4-treated sample compared with samples from other treatments.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization, LiChropur
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrochloric acid, puriss., 24.5-26.0%
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Supelco
Hydrogen chloride - 1-butanol solution, ~3 M in 1-butanol, for GC derivatization, LiChropur