Merck
  • Development of a benzophenone and alkyne functionalised trehalose probe to study trehalose dimycolate binding proteins.
Development of a benzophenone and alkyne functionalised trehalose probe to study trehalose dimycolate binding proteins.
Organic & biomolecular chemistry (2012-12-22)
Ashna A Khan, Faustin Kamena, Mattie S M Timmer, Bridget L Stocker
ABSTRACT
Trehalose dimycolates (TDMs) are the most abundant glycolipids found in the cell wall of Mycobacterium tuberculosis (M. tb). TDMs play an important role in the pathogenesis of M. tb yet the only known receptor for TDM is the macrophage inducible C-type lectin (mincle). To understand more about the interaction of TDMs with immune cells, affinity based proteome profiling (AfBPP) can be used to determine receptors that bind TDMs. To this end, we present the synthesis of the first AfBPP-TDM probe and report on its ability to activate macrophages. By doing so, we establish that the AfBPP-TDM probe appears to be a suitable substrate for future proteomic profiling experiments.
Sigma-Aldrich
Benzophenone, ReagentPlus®, 99%
Sigma-Aldrich
Benzophenone, purified by sublimation, ≥99%
Supelco
Benzophenone, analytical standard
Supelco
Benzophenone, Pharmaceutical Secondary Standard; Certified Reference Material
Benzophenone, European Pharmacopoeia (EP) Reference Standard
Supelco
Melting point standard 47-49°C, analytical standard
Supelco
Mettler-Toledo Calibration substance ME 18870, Benzophenone, traceable to primary standards (LGC)