Merck
  • A novel polymerization of ultrathin sensitive imprinted film on surface plasmon resonance sensor.
A novel polymerization of ultrathin sensitive imprinted film on surface plasmon resonance sensor.
The Analyst (2012-08-18)
Jianwei Dong, Yuan Peng, Na Gao, Jialei Bai, Baoan Ning, Ming Liu, Zhixian Gao
ABSTRACT
A new surface-initiated polymerization based on pasting the initiator on a sensor chip surface was applied to prepare a malachite green (MG) imprinted ultrathin film on a surface plasmon resonance (SPR) sensor. First, the initiator (2,2-azoisobutyronitrile) was pasted on the gold surface using polyvinyl chloride (PVC). The initiator-covered gold chip was then soaked in a pre-polymerization solution prepared by dissolving methacrylic acid (functional monomer), ethylene glycol dimethacrylate (cross-linker), and MG (template) in DMSO in a weighing bottle. Finally, the weighing bottle was placed in a vacuum oven and thermal-initiated polymerization was conducted at 60 °C for 16 h. This method was simple and time-saving compared with the commonly used surface-initiated polymerization. More importantly, the molecularly imprinted polymer (MIP) film prepared using this method was much thicker than that of commonly used methods; the adsorption quantity was also much larger. The MIP modified SPR sensor showed high sensitivity and selectivity as well as good stability in detecting MG. The results suggest that the ultrathin MIP film prepared using the new method in this study is suitable to serve as the recognition element of the SPR sensor.
Sigma-Aldrich
Schaeffer and Fulton Spore Stain Solution A, for microscopy
Sigma-Aldrich
Malachite Green oxalate salt, certified by the Biological Stain Commission
Sigma-Aldrich
Malachite Green oxalate salt, Technical grade
Supelco
Malachite Green oxalate salt, VETRANAL®, analytical standard
Sigma-Aldrich
Malachite Green oxalate salt, for microscopy, crystalline, S. No.: 754
Sigma-Aldrich
Malachite Green solution, for microscopy