Merck
  • A versatile method for functionalization and grafting of 2-hydroxyethyl cellulose (HEC) via Click chemistry.
A versatile method for functionalization and grafting of 2-hydroxyethyl cellulose (HEC) via Click chemistry.
Carbohydrate polymers (2012-07-31)
Ahmed M Eissa, Ezat Khosravi, A Levent Cimecioglu
ABSTRACT
This article describes a versatile method for the modification of 2-hydroxyethyl cellulose (HEC) involving azide-alkyne cycloaddition reaction to impart neutral (ester) and ionic (carboxylic acid and 1(ry) amine) functionalities. The synthetic approach involved, first the introduction of the azide functionality to HEC and then followed by its cycloaddition reaction with several alkyne terminated compounds: namely ethyl propiolate, 5-hexynoic acid and propargyl amine. Sequential Click reactions were also demonstrated to be feasible by the successful synthesis of polydimethylsiloxane (PDMS) grafted HEC containing neutral (ester) and ionic (carboxylic acid and 1(ry) amine) functionalities. The Click chemistry was then further utilized similarly to graft poly(lactic acid) (PLA) and poly(ethylene glycol) (PEG) segments to HEC to access its hydrophobic and hydrophilic analogs, respectively. AFM analysis revealed that while HEC itself formed uniform oval features, the PLA grafted HEC exhibited a brushlike architecture. The formation of these brushlike structures suggested that the HEC backbone exhibits an extended conformation with the side chains stretched out. The resulting polymeric materials were characterized by solution and solid state (13)C NMR and FTIR spectroscopy.
Sigma-Aldrich
2-Hydroxyethyl cellulose, average Mv ~720,000
Sigma-Aldrich
2-Hydroxyethyl cellulose, average Mv ~1,300,000
Sigma-Aldrich
2-Hydroxyethyl cellulose, average Mv ~90,000
Sigma-Aldrich
2-Hydroxyethyl cellulose, average Mw ~380,000
Sigma-Aldrich
Hydroxyethyl-cellulose, viscosity 90-160 cP, 5 % in H2O(25 °C)
Sigma-Aldrich
Hydroxyethyl-cellulose