Merck
Enhanced production of 2,3-butanediol by engineered Bacillus subtilis.
Applied microbiology and biotechnology (2012-03-01)
Ranjita Biswas, Masaru Yamaoka, Hideki Nakayama, Takashi Kondo, Ken-ichi Yoshida, Virendra S Bisaria, Akihiko Kondo
ABSTRACT
Production of 2,3-butanediol by Bacillus subtilis takes place in late-log or stationary phase, depending on the expression of bdhA gene encoding acetoin reductase, which converts acetoin to 2,3-butanediol. The present work focuses on the development of a strain of B. subtilis for enhanced production of 2,3-butanediol in early log phase of growth cycle. For this, the bdhA gene was expressed under the control of P( alsSD ) promoter of AlsSD operon for acetoin fermentation which served the substrate for 2,3-butanediol production. Addition of acetic acid in the medium induced the production of 2,3-butanediol by 2-fold. Two-step aerobic-anaerobic fermentation further enhanced 2,3-butanediol production by 4-fold in comparison to the control parental strain. Thus, addition of acetic acid and low dissolved oxygen in the medium are involved in activation of bdhA gene expression from P( alsSD ) promoter in early log phase. Under the conditions tested in this work, the maximum production of 2,3-butanediol, 2.1 g/l from 10 g/l glucose, was obtained at 24 h. Furthermore, under the optimized microaerophilic condition, the production of 2,3-butanediol improved up to 6.1 g/l and overall productivity increased by 6.7-fold to 0.4 g/l h in the engineered strain compared to that in the parental control.
Sigma-Aldrich
Acetoin, natural, ≥95%, FG
Sigma-Aldrich
Acetoin, May exist as crystalline dimer
Sigma-Aldrich
Acetoin, primarily dimer, ≥95%, FG
Supelco
Acetoin, analytical standard