Merck
  • pH-Induced molten globule state of Rhizopus niveus lipase is more resistant against thermal and chemical denaturation than its native state.
pH-Induced molten globule state of Rhizopus niveus lipase is more resistant against thermal and chemical denaturation than its native state.
Cell biochemistry and biophysics (2012-01-05)
Gulam Rabbani, Ejaz Ahmad, Nida Zaidi, Sadaf Fatima, Rizwan Hasan Khan
ABSTRACT
Here, we have characterized four pH-dependent states: alkaline state, "B" (pH 9.0), native state, "N" (pH 7.4), acid-induced state, "A" (pH 2.2) and molten globule state, "MG" (pH 1.8) of Rhizopus niveus lipase (RNL) by CD, tryptophanyl fluorescence, ANS binding, DLS, and enzyme activity assay. This "MG" state lacks catalytic activity and tertiary structure but it has native-like significant secondary structure. The "R (h)" of all the four states of RNL obtained from DLS study suggests that the molecular compactness of the protein increases as the pH of solution decreases. Kinetic analysis of RNL shows that it has maximum catalytic efficiency at state "B" which is 15-fold higher than state "N." The CD and tryptophanyl fluorescence studies of RNL on GuHCl and temperature-induced unfolding reveal that the "MG" state is more stable than the other states. The DSC endotherms of RNL obtained at pH 9.0, 7.4, and 2.2 were with two transitions, while at pH 1.8 it showed only a single transition.
Sigma-Aldrich
8-Anilino-1-naphthalenesulfonic acid ammonium salt, for fluorescence, ≥97.0% (HPLC)
Sigma-Aldrich
8-Anilino-1-naphthalenesulfonic acid
Sigma-Aldrich
8-Anilino-1-naphthalenesulfonic acid ammonium salt, technical, ≥90% (NT)
Sigma-Aldrich
8-Anilino-1-naphthalenesulfonic acid hemimagnesium salt hydrate, for fluorescence, ≥95.0% (T)
Sigma-Aldrich
8-Anilino-1-naphthalenesulfonic acid hemimagnesium salt hydrate, for fluorescence, ≥95% (TLC)