Merck
  • Multicomponent approaches to 8-carboxylnaphthyl-functionalized pyrazolo[3,4-b]pyridine derivatives.
Multicomponent approaches to 8-carboxylnaphthyl-functionalized pyrazolo[3,4-b]pyridine derivatives.
Organic & biomolecular chemistry (2011-12-15)
Yan Hao, Xiao-Ping Xu, Tao Chen, Li-Li Zhao, Shun-Jun Ji
ABSTRACT
A simple and novel protocol for the efficient synthesis of a series of 8-carboxylnaphthyl functionalized pyrazolo[3,4-b]pyridine derivatives was developed through a one-pot, three-component reaction involving acenaphthylene-1,2-dione and 1H-pyrazol-5-amine in acetic acid medium. The reaction represents the first facile conversion of acenaphthenequinone to naphthoic acid via C-C bond cleavage without need for multi-step transformation.
Product Description
Supelco
Acenaphthylene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Acenaphthylene, 75%
Supelco
Acenaphthylene, analytical standard
Sigma-Aldrich
Acenaphthylene, 99%