Merck
  • Liquid chromatography-mass spectrometry method for the quantification of tamoxifen and its metabolite 4-hydroxytamoxifen in rat plasma: application to interaction study with biochanin A (an isoflavone).
Liquid chromatography-mass spectrometry method for the quantification of tamoxifen and its metabolite 4-hydroxytamoxifen in rat plasma: application to interaction study with biochanin A (an isoflavone).
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2011-09-06)
Sheelendra Pratap Singh, Wahajuddin, Mushir M Ali, Kanchan Kohli, Girish Kumar Jain
ABSTRACT
Tamoxifen is the agent of choice for the treatment of estrogen receptor-positive breast cancer. Tamoxifen is a substrate of P-glycoprotein (P-gp) and microsomal cytochrome P450 (CYP) 3A, and biochanin A (BCA) is an inhibitor of P-gp and CYP3A. Hence, it could be expected that BCA would affect the pharmacokinetics of tamoxifen. In the present study we have developed and validated a simple, sensitive and specific LC-ESI-MS/MS method for the simultaneous quantification of tamoxifen and its metabolite 4-hydroxytamoxifen with 100 μL rat plasma using centchroman as an internal standard (IS). Tamoxifen, 4-hydroxytamoxifen and IS were separated on a Supelco Discovery C18 (4.6 mm × 50 mm, 5.0 μm) column under isocratic condition using 0.0 1M ammonium acetate (pH 4.5):acetonitrile (10:90, v/v) as a mobile phase. The mobile phase was delivered at a flow rate of 0.8 mL/min. The method was proved to be accurate and precise at linearity range of 0.78-200 ng/mL with a correlation coefficient (r) of ≥ 0.996. The intra- and inter-day assay precision ranged from 1.89 to 8.54% and 3.97 to 10.26%, respectively; and intra- and inter-day assay accuracy was between 87.63 and 109.06% and 96 and 103.89%, respectively for both the analytes. The method was successfully applied to study the effect of oral co-administration of BCA (an isoflavone) on the pharmacokinetics of tamoxifen and 4-hydroxytamoxifen in female rats. The coadministration of BCA caused no significant changes in the pharmacokinetics of tamoxifen and 4-hydroxytamoxifen. However, the peak plasma concentration (C(max)) of 4-hydroxytamoxifen in BCA pretreated rats was significantly (P<0.05) lower than those from control group.
Product Description
Supelco
Discovery® C18 Supelguard Guard Cartridge, Kit, 5 μm particle size, L × I.D. 2 cm × 4 mm
Supelco
Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 10 mm
Supelco
Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 2.1 mm
Supelco
Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 10 cm × 4 mm
Supelco
Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
Discovery® C18 Supelguard Guard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm, pkg of 2 ea
Supelco
Discovery® C18 Supelguard Guard Cartridge, Kit, 5 μm particle size, L × I.D. 2 cm × 2.1 mm
Supelco
Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 5 cm × 4.6 mm
Supelco
Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 10 cm × 4.6 mm
Supelco
Discovery® C18 Supelguard Guard Cartridge, 5 μm particle size, L × I.D. 2 cm × 2.1 mm, pkg of 2 ea
Supelco
Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 5 cm × 2.1 mm
Supelco
Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 21.2 mm
Supelco
Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 10 cm × 2.1 mm
Sigma-Aldrich
Biochanin A
Supelco
Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4 mm
Supelco
Discovery® C18 Supelguard Guard Cartridge, Kit, 5 μm particle size, L × I.D. 2 cm × 3 mm
Supelco
Discovery® C18 Supelguard Guard Cartridge, 5 μm particle size, L × I.D. 2 cm × 3 mm, pkg of 2 ea
Supelco
Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 12.5 cm × 2.1 mm
Supelco
Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 12.5 cm × 4 mm
Supelco
Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 3 mm
Supelco
Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4 mm
Supelco
Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 3 mm
Supelco
Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 12.5 cm × 4.6 mm