Merck
  • Renewable H2 from glycerol steam reforming: effect of La2O3 and CeO2 addition to Pt/Al2O3 catalysts.
Renewable H2 from glycerol steam reforming: effect of La2O3 and CeO2 addition to Pt/Al2O3 catalysts.
ChemSusChem (2010-04-28)
Tiziano Montini, Rakesh Singh, Piyali Das, Barbara Lorenzut, Nicolás Bertero, Pietro Riello, Alvise Benedetti, Giuliano Giambastiani, Claudio Bianchini, Sergey Zinoviev, Stanislav Miertus, Paolo Fornasiero
ABSTRACT
Glycerol is the main byproduct of biodiesel production and its increased production volume derives from the increasing demand for biofuels. The conversion of glycerol to hydrogen-rich mixtures presents an attractive route towards sustainable biodiesel production. Here we explored the use of Pt/Al(2)O(3)-based catalysts for the catalytic steam reforming of glycerol, evidencing the influence of La(2)O(3) and CeO(2) doping on the catalyst activity and selectivity. The addition of the latter metal oxides to a Pt/Al(2)O(3) catalyst is found to significantly improve the glycerol steam reforming, with high H(2) and CO(2) selectivities. A good catalytic stability is achieved for the Pt/La(2)O(3)/Al(2)O(3) system working at 350 degrees C, while the Pt/CeO(2)/Al(2)O(3) catalyst sharply deactivates after 20 h under similar conditions. Studies carried out on fresh and exhausted catalysts reveal that both systems maintain high surface areas and high Pt dispersions. Therefore, the observed catalyst deactivation can be attributed to coke deposition on the active sites throughout the catalytic process and only marginally to Pt nanoparticle sintering. This work suggests that an appropriate support composition is mandatory for preparing high-performance Pt-based catalysts for the sustainable conversion of glycerol into syngas.
Sigma-Aldrich
Lanthanum(III) oxide, 99.999% trace metals basis
Supelco
Lanthanum(III) oxide, suitable for AAS, ≥99.9%
Sigma-Aldrich
Lanthanum(III) oxide, ≥99.9%
Sigma-Aldrich
Lanthanum(III) oxide, 99.99% trace metals basis
Sigma-Aldrich
Lanthanum(III) oxide, nanopowder, <100 nm particle size (TEM), 99% trace metals basis