Merck
Synthesis of CuInSe(2) nanocrystals with trigonal pyramidal shape.
Journal of the American Chemical Society (2009-02-17)
Bonil Koo, Reken N Patel, Brian A Korgel
ABSTRACT
The synthesis of monodisperse chalcopyrite (tetragonal) CuInSe(2) nanocrystals is reported. The nanocrystals have trigonal pyramidal shape, and they exhibited a common crystallographic orientation when drop-cast onto carbon substrates. A crystallographic model for the nanocrystals was developed. The nanocrystals are bounded by one polar {112} surface facet and three nonpolar {114} surface facets.
Sigma-Aldrich
Copper(I) Chloride ChemBeads, 15 wt% loading
Sigma-Aldrich
Selenourea, 98%
Sigma-Aldrich
Copper(I) chloride, ≥99.995% trace metals basis
Sigma-Aldrich
Copper(I) chloride, ReagentPlus®, purified, ≥99%
Sigma-Aldrich
Copper(I) chloride, reagent grade, 97%
Sigma-Aldrich
Copper(I) chloride, AnhydroBeads, ≥99.99% trace metals basis
Sigma-Aldrich
Indium(III) chloride, anhydrous, powder, ≥99.999% trace metals basis
Sigma-Aldrich
Indium(III) chloride, 98%
Sigma-Aldrich
Indium(III) chloride, 99.999% trace metals basis