Merck
A presentation of longstanding toxoplasmosis chorioretinitis.
Optometry (St. Louis, Mo.) (2008-12-30)
Brian R Hall, Gary E Oliver, Mitchell Wilkinson
ABSTRACT
Toxoplasmosis gondii is the most common cause of focal necrotizing retinitis in healthy individuals. This case report describes a presentation of toxoplasmosis chorioretintis and reviews the current management options. A 10-year-old Hispanic girl presented with complaints of decreased vision in her right eye for 3 weeks. The patient had presumed ocular toxoplasmosis chorioretinitis with secondary granulomatous panuveitis. She was treated successfully with Bactrim (Roche Laboratories, Nutley, New Jersey) and topical steroids and cylcoplegics. Ocular toxoplasmosis is a self-limiting disease in immunocompetent individuals; however, proper diagnosis and early intervention improves visual outcome.
Sigma-Aldrich
Pheniramine maleate salt
Sigma-Aldrich
Naphazoline hydrochloride
Supelco
Pheniramine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®