Merck
  • Histamine H1-receptor blockade in humans affects psychomotor performance but not memory.
Histamine H1-receptor blockade in humans affects psychomotor performance but not memory.
Journal of psychopharmacology (Oxford, England) (2008-01-23)
P van Ruitenbeek, A Vermeeren, Wj Riedel
ABSTRACT
Results from recent animal studies suggest an important role for histamine in memory functioning. Histaminergic drugs might prove beneficial for people suffering from memory impairment. To determine if histamine is involved in memory functioning this study evaluates the effects of histaminergic dysfunction on memory performance by administrating a H1-antagonist to humans. The study was conducted according to a 4-way, double-blind, crossover design in 20 healthy female volunteers, aged 18-45 years. On each test day subjects completed three test sessions: before and around 2 and 4 h after administration of single oral doses of dexchlorpheniramine 2 mg or 4 mg, scopolamine 1 mg or placebo. Drug effects were assessed using tests of memory, psychomotor and attention performance, and subjective alertness. Results showed that dexchlorpheniramine impaired performance in tests of spatial learning, reaction time, tracking and divided attention but showed no effects on working memory, visual memory, word learning or memory scanning. Scopolamine induced a similar pattern of effects. In addition, both drugs decreased subjective alertness. In conclusion results show that dexchlorpheniramine and scopolamine clearly impaired performance on psychomotor and attention tasks but do not suggest a specific role of the histaminergic system in learning and memory in humans.
Product Description
Supelco
Chlorpheniramine Maleate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
S-(+)-Chlorpheniramine maleate salt
Dexchlorpheniramine maleate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
(±)-Chlorpheniramine maleate salt, ≥99% (perchloric acid titration)
Chlorphenamine maleate, European Pharmacopoeia (EP) Reference Standard
Supelco
Chlorpheniramine maleate salt solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®