Merck
  • Effects of slow-release urea fertilizers on urease activity, microbial biomass, and nematode communities in an aquic brown soil.
Effects of slow-release urea fertilizers on urease activity, microbial biomass, and nematode communities in an aquic brown soil.
Science in China. Series C, Life sciences (2005-08-11)
Xiaoguang Jiao, Wenju Liang, Lijun Chen, Haijun Zhang, Qi Li, Peng Wang, Dazhong Wen
ABSTRACT
A field experiment was carried out at the Shenyang Experimental Station of Ecology (CAS) in order to study the effects of slow-release urea fertilizers high polymer-coated urea (SRU1), SRU1 mixed with dicyandiamide DCD (SRU2), and SRU1 mixed with calcium carbide CaC2 (SRU3) on urease activity, microbial biomass C and N, and nematode communities in an aquic brown soil during the maize growth period. The results demonstrated that the application of slow-release urea fertilizers inhibits soil urease activity and increases the soil NH4+-N content. Soil available N increment could promote its immobilization by microorganisms. Determination of soil microbial biomass N indicated that a combined application of coated urea and nitrification inhibitors increased the soil active N pool. The population of predators/omnivores indicated that treatment with SRU2 could provide enough soil NH4+-N to promote maize growth and increased the food resource for the soil fauna compared with the other treatments.
Sigma-Aldrich
Calcium carbide, pieces, thickness <10 mm , typically, technical grade, ~80%
Sigma-Aldrich
Calcium carbide, granulated, technical, ≥75% (gas-volumetric)