Merck
  • Important aroma compounds in freshly ground wholemeal and white wheat flour-identification and quantitative changes during sourdough fermentation.
Important aroma compounds in freshly ground wholemeal and white wheat flour-identification and quantitative changes during sourdough fermentation.
Journal of agricultural and food chemistry (2002-10-31)
Michael Czerny, Peter Schieberle
ABSTRACT
An investigation on the odor-active compounds of wholemeal (WWF) and white wheat flour (WF 550) by aroma extract dilution analysis (AEDA) and by quantitative studies using stable isotope dilution assays (SIDA) revealed a significant number of odor-active compounds, such as (E)-2-nonenal, (E,Z)- and (E,E)-2,4-decadienal, (E)-4,5-epoxy-(E)-2-decenal, 3-hydroxy-4,5-dimethyl-2(5H)-furanone, and vanillin, with high odor activities in both wheat flours. The amounts and, consequently, the aroma potencies of vanillin, (E,E)-2,4-decadienal, and 3-(methylthio)propanal were much higher in the WWF than in the WF 550 samples. Fermentation of suspensions of both flours with lactic acid bacteria did not generate new odorants; however, many compounds, such as acetic acid or 3-methylbutanal, were increased, whereas aldehydes (formed from the degradation of unsaturated fatty acids) were decreased. Comparing the odorant concentrations present before and after fermentation gave evidence that the main influence of the microorganisms on sourdough aroma is to either enhance or decrease specific volatiles already present in the flour. A comparison with literature data indicated that most of these odorants are also important for the bread crumb aroma present after baking of the dough.
Sigma-Aldrich
trans,trans-2,6-Nonadienal, ≥95%, stabilized, FG
Sigma-Aldrich
trans-2,cis-6-Nonadienal, 95%
Sigma-Aldrich
trans-2,cis-6-Nonadienal, mixture of isomers, ≥96%, stabilized, FCC, FG