Merck
  • Low-abundant protein extraction from complex protein sample using a novel continuous aqueous two-phase systems device.
Low-abundant protein extraction from complex protein sample using a novel continuous aqueous two-phase systems device.
Journal of separation science (2012-12-21)
Patricia Vázquez-Villegas, Edith Espitia-Saloma, Marco Rito-Palomares, Oscar Aguilar
ABSTRACT
The present work describes the application of a novel continuous aqueous two-phase system prototype for the recovery of biomolecules. The prototype is an alternative platform for protein recovery and α-amylase from soybean extracts was used as a model system. The system was selected as an example of low-abundant protein present in complex mixtures. Compared with batch systems, continuous operation in this prototype seems to increase partition coefficient with higher recovery efficiencies. Processing time is reduced at least three times in the continuous system when compared to batch mode, while hold up (volumetric quantity of the opposing phase in a determined phase sample) decreases with decreasing phases flow. Furthermore, similar partition coefficient (Kp > 4) with a higher top phase enzyme recovery (81%) is also obtained in this system probably due to better contact surface between phases, compared with that obtained in batch (79%). A continuous aqueous two-phase system process with purification factor 40-fold higher than batch experiments was achieved. These preliminary results exhibit the potential of continuous systems for the recovery of low-abundant proteins from complex mixtures. The promising performance of this prototype can raise the attention of the industry for the adoption of aqueous two-phase system processes.
Sigma-Aldrich
Taka-Diastase from Aspergillus oryzae, powder, slightly beige, ~100 U/mg
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, yellow-brown, ~380 U/mg
Sigma-Aldrich
α-Amylase from porcine pancreas, Type VI-B, ≥5 units/mg solid
Sigma-Aldrich
α-Amylase from porcine pancreas, Type I-A, PMSF treated, saline suspension, 700-1400 units/mg protein (E1%/280)
Sigma-Aldrich
α-Amylase from Bacillus sp., Type II-A, lyophilized powder, ≥1,500 units/mg protein (biuret)
Sigma-Aldrich
α-Amylase, heat-stable, solution, for use in Total Dietary Fiber Assay, TDF-100A
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, ≥400 units/mg protein (Lowry)
Sigma-Aldrich
α-Amylase from porcine pancreas, PMSF Treated, Type I-A, saline suspension, ≥1000 units/mg protein (E1%/280)
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, yellow-brown, ~50 U/mg
Sigma-Aldrich
α-Amylase from Bacillus sp., liquid
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, powder, ~1.5 U/mg (~0.2 U acc. to Willstätter)
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, aqueous solution, ≥800 FAU/g
Supelco
α-Amylase from Bacillus licheniformis, suitable for determination of starch (Kit STA-20)
Sigma-Aldrich
α-Amylase from human saliva, Type XIII-A, lyophilized powder, 300-1,500 units/mg protein
Sigma-Aldrich
α-Amylase from Bacillus licheniformis, Type XII-A, saline solution, ≥500 units/mg protein (biuret)
Sigma-Aldrich
α-Amylase from human saliva, Type IX-A, lyophilized powder, 1,000-3,000 units/mg protein
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, ≥150 units/mg protein (biuret)
Sigma-Aldrich
α-Amylase from Bacillus amyloliquefaciens, liquid, ≥250 units/g
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, powder, ~30 U/mg
Sigma-Aldrich
α-Amylase from Bacillus licheniformis, lyophilized powder, 500-1,500 units/mg protein, 93-100% (SDS-PAGE)