Merck
  • Inhibition of Amino Acids Influx into Proximal Tubular Cells Improves Lysosome Function in Diabetes.
Inhibition of Amino Acids Influx into Proximal Tubular Cells Improves Lysosome Function in Diabetes.
Kidney360 (2023-12-08)
Yuzuki Kano, Satoshi Yamaguchi, Koki Mise, Chieko Kawakita, Yasuhiro Onishi, Naoko Kurooka, Ryosuke Sugawara, Haya Hamed Hassan Albuayjan, Atsuko Nakatsuka, Jun Eguchi, Jun Wada
ABSTRACT
Collectrin serves as a chaperone for the trafficking of neutral amino acid (AA) transporters in the apical membranes of proximal tubular cells (PTCs). Cltrn knockout reduced AAs influx into PTCs, inactivated mTOR, activated transcription factor EB, improved lysosome function, and ameliorated vacuolar formation of PTCs in diabetic mice treated with streptozotocin and high-fat diet. The inhibition of neutral AA transporter, such as B0AT1 (SLC6A19), and transcription factor EB activator is a new therapeutic strategy against diabetic kidney disease. Inhibition of glucose influx into proximal tubular cells (PTCs) by sodium–glucose cotransporter 2 inhibitors revealed prominent therapeutic effects on diabetic kidney disease. Collectrin (CLTRN) serves as a chaperone for the trafficking of neutral amino acid (AA) transporters in the apical membranes of PTCs. We investigated the beneficial effects of reduced influx of AAs into PTCs in diabetes and obesity model of Cltrn−/y mice. Cltrn+/y and Cltrn−/y mice at age 5 weeks were assigned to standard diet and streptozotocin and high-fat diet (STZ-HFD)–treated groups. At age 22–23 weeks, body weight and HbA1c levels significantly increased in STZ-HFD-Cltrn+/y compared with standard diet-Cltrn+/y; however, they were not altered in STZ-HFD-Cltrn−/y compared with STZ-HFD-Cltrn+/y. At age 20 weeks, urinary albumin creatinine ratio was significantly reduced in STZ-HFD-Cltrn−/y compared with STZ-HFD-Cltrn+/y. Under the treatments with STZ and HFD, the Cltrn gene deficiency caused significant increase in urinary concentration of AAs such as Gln, His, Gly, Thr, Tyr, Val, Trp, Phe, Ile, Leu, and Pro. In PTCs in STZ-HFD-Cltrn+/y, the enlarged lysosomes with diameter of 10 μm or more were associated with reduced autolysosomes, and the formation of giant lysosomes was prominently suppressed in STZ-HFD-Cltrn−/y. Phospho-mTOR and inactive form of phospho-transcription factor EB were reduced in STZ-HFD-Cltrn−/y compared with STZ-HFD-Cltrn+/y. The reduction of AAs influx into PTCs inactivated mTOR, activated transcription factor EB, improved lysosome function, and ameliorated vacuolar formation of PTCs in STZ-HFD-Cltrn−/y mice.