- Synthesis of an antisense oligonucleotide targeted against C-raf kinase: efficient oligonucleotide synthesis without chlorinated solvents.
Synthesis of an antisense oligonucleotide targeted against C-raf kinase: efficient oligonucleotide synthesis without chlorinated solvents.
Bioorganic & medicinal chemistry (1999-04-29)
A H Krotz, D L Cole, V T Ravikumar
PMID10220029
ABSTRACT
It is demonstrated that a solution of dichloroacetic acid in toluene removes dimethoxytrityl groups from the 5'-terminus of an antisense phosphorothioate oligodeoxyribonucleotide (ISIS 5132/CGP69846A) during synthesis on solid support cleanly and efficiently. It is therefore suggested to replace health hazardous dichloromethane which is typically used in oligonucleotide synthesis as solvent for DMTr-removal by toluene.