Merck
  • Surface/Interfacial Structure and Chemistry of High-Energy Nickel-Rich Layered Oxide Cathodes: Advances and Perspectives.
Surface/Interfacial Structure and Chemistry of High-Energy Nickel-Rich Layered Oxide Cathodes: Advances and Perspectives.
Small (Weinheim an der Bergstrasse, Germany) (2017-10-05)
Peiyu Hou, Jiangmei Yin, Meng Ding, Jinzhao Huang, Xijin Xu
ABSTRACT
The urgent prerequisites of high energy-density and superior electrochemical properties have been the main inspiration for the advancement of cathode materials in lithium-ion batteries (LIBs) in the last two decades. Nickel-rich layered transition-metal oxides with large reversible capacity as well as high operating voltage are considered as the most promising candidate for next-generation LIBs. Nonetheless, the poor long-term cycle-life and inferior thermal stability have limited their broadly practical applications. In the research of LIBs, it is observed that surface/interfacial structure and chemistry play significant roles in the performance of cathode cycling. This is due to the fact that they are basically responsible for the reversibility of Li+ intercalation/deintercalation chemistries while dictating the kinetics of the general cell reactions. In this Review, the surface/interfacial structure and chemistry of nickel-rich layered cathodes involving structural defects, redox mechanisms, structural evolutions, side-reactions among others are initially demonstrated. Recent advancements in stabilizing the surface/interfacial structure and chemistry of nickel-rich cathodes by surface modification, core-shell/concentration-gradient structure, foreign-ion substitution, hybrid surface, and electrolyte additive are presented. Then lastly, the remaining challenges such as the fundamental studies and commercialized applications, as well as the future research directions are discussed.
Sigma-Aldrich
NMC532, electrode sheet, aluminum substrate, size 5 in. × 10 in.
Sigma-Aldrich
NMC622, electrode sheet, aluminum substrate, size 5 in. × 10 in.
Sigma-Aldrich
NMC811, electrode sheet, aluminum substrate, size 5 in. × 10 in.