Merck
  • TGF-Beta Induces Activin A Production in Dermal Fibroblasts Derived from Patients with Fibrodysplasia Ossificans Progressiva.
TGF-Beta Induces Activin A Production in Dermal Fibroblasts Derived from Patients with Fibrodysplasia Ossificans Progressiva.
International journal of molecular sciences (2023-02-12)
Ruben D de Ruiter, Lisanne E Wisse, Ton Schoenmaker, Maqsood Yaqub, Gonzalo Sánchez-Duffhues, E Marelise W Eekhoff, Dimitra Micha
ABSTRACT
Fibrodysplasia ossificans progressiva (FOP) is a catastrophic, ultra-rare disease of heterotopic ossification caused by genetic defects in the ACVR1 gene. The mutant ACVR1 receptor, when triggered by an inflammatory process, leads to heterotopic ossification of the muscles and ligaments. Activin A has been discovered as the main osteogenic ligand of the FOP ACVR1 receptor. However, the source of Activin A itself and the trigger of its production in FOP individuals have remained elusive. We used primary dermal fibroblasts from five FOP patients to investigate Activin A production and how this is influenced by inflammatory cytokines in FOP. FOP fibroblasts showed elevated Activin A production compared to healthy controls, both in standard culture and osteogenic transdifferentiation conditions. We discovered TGFβ1 to be an FOP-specific stimulant of Activin A, shown by the upregulation of the INHBA gene and protein expression. Activin A and TGFβ1 were both induced by BMP4 in FOP and control fibroblasts. Treatment with TNFα and IL6 produced negligible levels of Activin A and TGFβ1 in both cell groups. We present for the first time TGFβ1 as a triggering factor of Activin A production in FOP. As TGFβ1 can promote the induction of the main driver of FOP, TGFβ1 could also be considered a possible therapeutic target in FOP treatment.
Sigma-Aldrich
Activin A human, ≥95% (SDS-PAGE), recombinant, expressed in baculovirus infected insect cells, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Anti-phospho Smad1/Smad5/Smad8 (Ser463/465), from rabbit, purified by affinity chromatography