Merck
Axonal components of nerves innervating the human arm.
Annals of neurology (2017-08-24)
Bernhard Gesslbauer, Laura A Hruby, Aidan D Roche, Dario Farina, Roland Blumer, Oskar C Aszmann
ABSTRACT
Axons traveling within the brachial plexus are responsible for the dexterous control of human arm and hand movements. Despite comprehensive knowledge on the topographical anatomy of nerves innervating the human upper limbs, the definite quantity of sensory and motor axons within this neural network remains elusive. Our aim was to perform a quantitative analysis of the axonal components of human upper limb nerves based on highly specific molecular features from spinal cord level to the terminal nerves at wrist level. Nerve specimen harvest at predefined harvesting sites (plexus roots and cords as well as major nerves originating from the brachial plexus innervating the arm and hand) was performed in 9 human heart-beating organ donors. Double immunofluorescence staining using antibodies against choline-acetyltransferase and neurofilament was performed to differentiate motor and sensory axons on nerve cross sections. Three hundred fifty thousand axons emerge from the spinal cord to innervate the human upper limb, of which 10% are motor neurons. In all nerves studied, sensory axons outnumber motor axons by a ratio of at least 9:1. The sensory axon contribution increases when moving distally, whereas only 1,700 motor axons reach the hand to innervate the intrinsic musculature. Our results suggest that upper limb motor execution, and particularly dexterous coordination of hand movement, require an unexpectedly low number of motor neurons, with a large convergence of afferent input for feedback control. Ann Neurol 2017;82:396-408.
Sigma-Aldrich
Anti-Choline Acetyltransferase Antibody, Chemicon®, from goat
Sigma-Aldrich
Anti-Neurofilament H Antibody, Chemicon®, from chicken