Merck
Derivation of neural crest cells from human pluripotent stem cells.
Nature protocols (2010-04-03)
Gabsang Lee, Stuart M Chambers, Mark J Tomishima, Lorenz Studer
ABSTRACT
Human pluripotent stem cell (hPSC)-derived neural crest (NC) cells present a valuable tool for modeling aspects of human NC development, including cell fate specification, multipotency and cell migration. hPSC-derived NC cells are also suitable for modeling human disease and as a renewable cell source for applications in regenerative medicine. Here we provide protocols for the step-wise differentiation of human embryonic stem cells (hESCs) or human induced pluripotent stem cells (hiPSCs) into neuroectodermal and NC cells using either the MS5 coculture system or a novel defined culture method based on pharmacological inhibition of bone morphogenetic protein and transforming growth factor-beta signaling pathways. Furthermore, we present protocols for the purification and propagation of hPSC-NC cells using flow cytometry and defined in vitro culture conditions. Our protocol has been validated in multiple independent hESC and hiPSC lines. The average time required for generating purified hPSC-NC precursors using this protocol is 2-5 weeks.
Sigma-Aldrich
Sodium selenite, γ-irradiated, lyophilized powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Gelatin from porcine skin, powder, gel strength ~300 g Bloom, Type A, BioReagent, suitable for electrophoresis, suitable for cell culture
Sigma-Aldrich
Anti-HNK-1/N-CAM (CD57) antibody, Mouse monoclonal, clone VC1.1, purified from hybridoma cell culture
Sigma-Aldrich
SU5402, SU5402, CAS 215543-92-3, is a cell-permeable, reversible, and ATP-competitive inhibitor of the tyrosine kinase activity of FGFR1 (IC₅₀ = 10-20 µM in the presence of 1 mM ATP).
Sigma-Aldrich
Anti-Brn-3a Antibody, Chemicon®, from rabbit
Sigma-Aldrich
N6,2′-O-Dibutyryladenosine 3′,5′-cyclic monophosphate sodium salt, ≥97% (HPLC), powder
Sigma-Aldrich
N-Acetyl-Lys-Pro-Arg-7-amido-4-trifluoromethylcoumarin, ≥95% (HPLC), solid