Merck
  • MiR-7015-3p Targets Nuclear Factor-Kappa-B-Inhibitor Alpha to Aggravate Hypoxia/Reoxygenation Injury in Cardiomyocytes Through the NF-κB Pathway.
MiR-7015-3p Targets Nuclear Factor-Kappa-B-Inhibitor Alpha to Aggravate Hypoxia/Reoxygenation Injury in Cardiomyocytes Through the NF-κB Pathway.
International heart journal (2022-10-03)
Xin Shen, Jing Tao, Zhao Wang, Guoqing Li, Zilong Zhang, Jie Li, Adri Diliar
ABSTRACT
Ischemic heart disease (IHD) is a prominent global cause of morbidity and death resulting from the narrowing or blockage of cardiac coronary arteries. Exposing isolated cardiac myocytes to hypoxia-reoxygenation (H/R) might be an efficient tool to investigate the etiology and underlying mechanism of myocardial ischemia-reperfusion (I/R) injury. This study found that miR-7015 is upregulated in mouse myocardial tissues after I/R injury and in cardiomyocytes after H/R injury. A model of H/R-induced cardiomyocyte injury was established; miR-7015 overexpression exacerbated while miR-7015 inhibition partially ameliorated H/R-induced cardiomyocyte injury by inhibiting cytokine release, promoting cell viability, and suppressing apoptosis. Bioinformatics and experimental studies have identified nuclear factor-kappa-B-inhibitor alpha (Nfkbia) as a direct downstream target of miR-7015. miR-7015 inhibited Nfkbia expression. Unlike miR-7015 overexpression, Nfkbia overexpression alleviated H/R-induced injury in cardiomyocytes. Moreover, Nfkbia overexpression partially abolished the effects of miR-7015 overexpression on H/R-induced cardiomyocyte injury. In conclusion, the miR-7015/Nfkbia axis modulates cardiomyocyte injury induced by H/R, possibly through the NF-κB signaling.
Sigma-Aldrich
Mouse IL-1 β ELISA Kit, for cell and tissue lysates
Sigma-Aldrich
Mouse Tumor Necrosis Factor α ELISA Kit, for serum, plasma and cell culture supernatant