Merck
  • UHPLC-electrospray ionization-mass spectrometric analysis of brain cell-specific glucogenic and neurotransmitter amino acid content.
UHPLC-electrospray ionization-mass spectrometric analysis of brain cell-specific glucogenic and neurotransmitter amino acid content.
Scientific reports (2021-08-11)
Khaggeswar Bheemanapally, Prabhat R Napit, Mostafa M H Ibrahim, Karen P Briski
ABSTRACT
Astrocyte glycogen, the primary energy reserve in brain, undergoes continuous remodeling by glucose passage through the glycogen shunt prior to conversion to the oxidizable energy fuel L-lactate. Glucogenic amino acids (GAAs) are a potential non-glucose energy source during neuro-metabolic instability. Current research investigated whether diminished glycogen metabolism affects GAA homeostasis in astrocyte and/or nerve cell compartments. The glycogen phosphorylase (GP) inhibitor 1,4-dideoxy-1,4-imino-D-arabinitol (DAB) was injected into the ventromedial hypothalamic nucleus (VMN), a key metabolic-sensing structure, before vehicle or L-lactate infusion. Pure VMN astrocyte and metabolic-sensory neuron samples were obtained by combinatory immunocytochemistry/laser-catapult-microdissection for UHPLC-electrospray ionization-mass spectrometry (LC-ESI-MS) GAA analysis. DAB inhibition of VMN astrocyte aspartate and glutamine (Gln) levels was prevented or exacerbated, respectively, by lactate. VMN gluco-stimulatory nitric oxide (NO; neuronal nitric oxide synthase-immunoreactive (ir)-positive) and gluco-inhibitory γ-aminobutyric acid (GABA; glutamate decarboxylase65/67-ir-positive) neurons exhibited lactate-reversible asparate and glutamate augmentation by DAB, but dissimilar Gln responses to DAB. GP inhibition elevated NO and GABA nerve cell GABA content, but diminished astrocyte GABA; these responses were averted by lactate in neuron, but not astrocyte samples. Outcomes provide proof-of-principle of requisite LC-ESI-MS sensitivity for GAA measurement in specific brain cell populations. Results document divergent effects of decreased VMN glycogen breakdown on astrocyte versus neuron GAAs excepting Gln. Lactate-reversible DAB up-regulation of metabolic-sensory neuron GABA signaling may reflect compensatory nerve cell energy stabilization upon decline in astrocyte-derived metabolic fuel.