Merck
  • Inverse agonism at the Na/K-ATPase receptor reverses EMT in prostate cancer cells.
Inverse agonism at the Na/K-ATPase receptor reverses EMT in prostate cancer cells.
The Prostate (2021-05-07)
Moumita Banerjee, Zhichuan Li, Yingnyu Gao, Fangfang Lai, Minqi Huang, Zhongbing Zhang, Liquan Cai, Juan Sanabria, Tianyan Gao, Zijian Xie, Sandrine V Pierre
ABSTRACT
The surface expression of Na/K-ATPase α1 (NKA) is significantly reduced in primary prostate tumors and further decreased in bone metastatic lesions. Here, we show that the loss of cell surface expression of NKA induces epithelial-mesenchymal transition (EMT) and promotes metastatic potential and tumor growth of prostate cancer (PCa) by decreasing the expression of E-cadherin and increasing c-Myc expression via the activation of Src/FAK pathways. Mechanistically, reduced surface expression of NKA in PCa is due to increased endocytosis through the activation of NKA/Src receptor complex. Using a high-throughput NKA ligand-screening platform, we have discovered MB5 as an inverse agonist of the NKA/Src receptor complex, capable of blocking the endocytosis of NKA. MB5 treatment increased NKA expression and E-cadherin in PCa cells, which reversed EMT and consequently decreased the invasion and growth of spheroid models and tumor xenografts. Thus, we have identified a hitherto unrecognized mechanism that regulates EMT and invasiveness of PCa and demonstrated for the first time the feasibility of identifying inverse agonists of receptor NKA/Src complex and their potential utility as anticancer drugs. We, therefore, conclude that cell surface expression of α1 NKA can be targeted for the development of new therapeutics against aggressive PCa and that MB5 may serve as a prototype for drug development against EMT in metastatic PCa.
Sigma-Aldrich
PP2, ≥98% (HPLC)
Sigma-Aldrich
Anti-Phosphotyrosine Antibody, clone 4G10®, clone 4G10®, Upstate®, from mouse
Sigma-Aldrich
Anti-Na+/K+ ATPase β-1 Antibody, clone C464.8, clone C464.8, Upstate®, from mouse
Sigma-Aldrich
Anti-Na+/K+ ATPase α-1 Antibody, clone C464.6, clone C464.6, Upstate®, from mouse