Merck
  • Cutaneous pigmentation modulates skin sensitivity via tyrosinase-dependent dopaminergic signalling.
Cutaneous pigmentation modulates skin sensitivity via tyrosinase-dependent dopaminergic signalling.
Scientific reports (2017-08-25)
Kentaro Ono, Chi T Viet, Yi Ye, Dongmin Dang, Suzuro Hitomi, Takashi Toyono, Kiyotoshi Inenaga, John C Dolan, Brian L Schmidt
ABSTRACT
We propose a new mechanism of sensory modulation through cutaneous dopaminergic signalling. We hypothesize that dopaminergic signalling contributes to differential cutaneous sensitivity in darker versus lighter pigmented humans and mouse strains. We show that thermal and mechanical cutaneous sensitivity is pigmentation dependent. Meta-analyses in humans and mice, along with our own mouse behavioural studies, reveal higher thermal sensitivity in pigmented skin relative to less-pigmented or albino skin. We show that dopamine from melanocytes activates the D1-like dopamine receptor on primary sensory neurons. Dopaminergic activation increases expression of the heat-sensitive TRPV1 ion channel and reduces expression of the mechanically-sensitive Piezo2 channel; thermal threshold is lower and mechanical threshold is higher in pigmented skin.
Sigma-Aldrich
Dopamine hydrochloride
Sigma-Aldrich
(−)-Norepinephrine, ≥98%, crystalline
Sigma-Aldrich
Anti-Dopamine D₅ Receptor Rabbit pAb, lyophilized, Calbiochem®
Sigma-Aldrich
(±)-Epinephrine hydrochloride