Merck
Transcriptomic Analysis in the Sea Anemone Nematostella vectensis.
Methods in molecular biology (Clifton, N.J.) (2020-10-20)
Jacob F Warner, Eric Röttinger
ABSTRACT
The sea anemone Nematostella vectensis is an emerging research model to study embryonic development and regeneration at the molecular and global transcriptomic level. Transcriptomics analysis is now routinely used to detect differential expression at the genome level. Here we present the latest procedures for isolating high-quality RNA required for next generation sequencing, as well as methods and resources for quantifying transcriptomic data.
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Phenol:Chloroform:Isoamyl Alcohol 25:24:1 Saturated with 10 mM Tris, pH 8.0, 1 mM EDTA, Supplied with Equilibration buffer