Merck
  • Fluorinated pyridinium oximes as potential reactivators for acetylcholinesterases inhibited by paraoxon organophosphorus agent.
Fluorinated pyridinium oximes as potential reactivators for acetylcholinesterases inhibited by paraoxon organophosphorus agent.
Bioorganic & medicinal chemistry (2009-08-12)
Hee Chun Jeong, No-Joong Park, Chong Hak Chae, Kamil Musilek, Jiri Kassa, Kamil Kuca, Young-Sik Jung
ABSTRACT
A series of fluorinated oxime compounds was designed and synthesized in order to probe the effect of fluorine substitution on reactivation of inhibited acetylcholinesterase (AChE) by organophosphorus agents. Permeability measurements, using the Parallel Artificial Membrane Permeation Assays (PAMPA) method, were employed to experimentally demonstrate that membrane permeabilities of the series of oximes increase in proportional to the increase in the number of fluorine atoms. Among the compounds explored in this study, the mono-fluorinated carbamoyl aldoxime 4b was the most potent reactivator for paraoxon-inhibited red blood cell (RBC) AChE.
Product Description
Supelco
Warfarin, PESTANAL®, analytical standard
Supelco
Warfarin, analytical standard
Millipore
Multiscreen® 96 well Plate, hydrophobic PVDF membrane, pore size 0.45 μm, non-sterile
Sigma-Aldrich
Pyridine-2-aldoxime methochloride