Merck
  • ISG15 and ISGylation is required for pancreatic cancer stem cell mitophagy and metabolic plasticity.
ISG15 and ISGylation is required for pancreatic cancer stem cell mitophagy and metabolic plasticity.
Nature communications (2020-05-31)
Sonia Alcalá, Patricia Sancho, Paola Martinelli, Diego Navarro, Coral Pedrero, Laura Martín-Hijano, Sandra Valle, Julie Earl, Macarena Rodríguez-Serrano, Laura Ruiz-Cañas, Katerin Rojas, Alfredo Carrato, Laura García-Bermejo, Miguel Ángel Fernández-Moreno, Patrick C Hermann, Bruno Sainz
ABSTRACT
Pancreatic cancer stem cells (PaCSCs) drive pancreatic cancer tumorigenesis, chemoresistance and metastasis. While eliminating this subpopulation of cells would theoretically result in tumor eradication, PaCSCs are extremely plastic and can successfully adapt to targeted therapies. In this study, we demonstrate that PaCSCs increase expression of interferon-stimulated gene 15 (ISG15) and protein ISGylation, which are essential for maintaining their metabolic plasticity. CRISPR-mediated ISG15 genomic editing reduces overall ISGylation, impairing PaCSCs self-renewal and their in vivo tumorigenic capacity. At the molecular level, ISG15 loss results in decreased mitochondrial ISGylation concomitant with increased accumulation of dysfunctional mitochondria, reduced oxidative phosphorylation (OXPHOS) and impaired mitophagy. Importantly, disruption in mitochondrial metabolism affects PaCSC metabolic plasticity, making them susceptible to prolonged inhibition with metformin in vivo. Thus, ISGylation is critical for optimal and efficient OXPHOS by ensuring the recycling of dysfunctional mitochondria, and when absent, a dysregulation in mitophagy occurs that negatively impacts PaCSC stemness.
Sigma-Aldrich
Acridine Orange 10-nonyl bromide
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium, Without glucose, L-glutamine, phenol red, sodium pyruvate and sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
Acridine Orange hydrochloride solution, 10 mg/mL in H2O, ≥95.0% (HPLC)
Sigma-Aldrich
Fumitremorgin C, from Neosartorya fischeri, film
Sigma-Aldrich
Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone, ≥98% (TLC), powder
Sigma-Aldrich
Cyclosporin A, from Tolypocladium inflatum, ≥95% (HPLC), solid
Roche
ATP Bioluminescence Assay Kit CLS II, sufficient for 800 assays (tubes), kit of 1 (2 components), suitable for detection