Merck
  • Wnt signaling contributes to withdrawal symptoms from opioid receptor activation induced by morphine exposure or chronic inflammation.
Wnt signaling contributes to withdrawal symptoms from opioid receptor activation induced by morphine exposure or chronic inflammation.
Pain (2019-11-19)
Mingzheng Wu, Zehua Li, Lei Liang, Pingchuan Ma, Dong Cui, Peng Chen, Genhao Wu, Xue-Jun Song
ABSTRACT
Preventing and treating opioid dependence and withdrawal is a major clinical challenge, and the underlying mechanisms of opioid dependence and withdrawal remain elusive. We hypothesized that prolonged morphine exposure or chronic inflammation-induced μ-opioid receptor activity serves as a severe stress that elicits neuronal alterations and recapitulates events during development. Here, we report that Wnt signaling, which is important in developmental processes of the nervous system, plays a critical role in withdrawal symptoms from opioid receptor activation in mice. Repeated exposures of morphine or peripheral inflammation produced by intraplantar injection of complete Freund's adjuvant significantly increase the expression of Wnt5b in the primary sensory neurons in dorsal root ganglion (DRG). Accumulated Wnt5b in DRG neurons quickly transmits to the spinal cord dorsal horn (DH) after naloxone treatment. In the DH, Wnt5b, acts through the atypical Wnt-Ryk receptor and alternative Wnt-YAP/TAZ signaling pathways, contributing to the naloxone-precipitated opioid withdrawal-like behavioral symptoms and hyperalgesia. Inhibition of Wnt synthesis and blockage of Wnt signaling pathways greatly suppress the behavioral and neurochemical alterations after naloxone-precipitated withdrawal. These findings reveal a critical mechanism underlying naloxone-precipitated opioid withdrawal, suggesting that targeting Wnt5b synthesis in DRG neurons and Wnt signaling in DH may be an effective approach for prevention and treatment of opioid withdrawal syndromes, as well as the transition from acute to chronic pain.
Sigma-Aldrich
Lectin from Bandeiraea simplicifolia (Griffonia simplicifolia), Isolectin B4 (BSI-B4), biotin conjugate, lyophilized powder
Sigma-Aldrich
Anti-Active-β-Catenin (Anti-ABC) Antibody, clone 8E7, clone 8E7, Upstate®, from mouse
Sigma-Aldrich
Anti-phospho NR1 (Ser897) Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-GAPDH antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Wnt3A Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse