Merck
  • Thymosin β4 is essential for adherens junction stability and epidermal planar cell polarity.
Thymosin β4 is essential for adherens junction stability and epidermal planar cell polarity.
Development (Cambridge, England) (2020-12-15)
Krishnanand Padmanabhan, Hanna Grobe, Jonathan Cohen, Arad Soffer, Adnan Mahly, Orit Adir, Ronen Zaidel-Bar, Chen Luxenburg
ABSTRACT
Planar cell polarity (PCP) is essential for tissue morphogenesis and homeostasis; however, the mechanisms that orchestrate the cell shape and packing dynamics required to establish PCP are poorly understood. Here, we identified a major role for the globular (G)-actin-binding protein thymosin-β4 (TMSB4X) in PCP establishment and cell adhesion in the developing epidermis. Depletion of Tmsb4x in mouse embryos hindered eyelid closure and hair-follicle angling owing to PCP defects. Tmsb4x depletion did not preclude epidermal cell adhesion in vivo or in vitro; however, it resulted in abnormal structural organization and stability of adherens junction (AJ) due to defects in filamentous (F)-actin and G-actin distribution. In cultured keratinocytes, TMSB4X depletion increased the perijunctional G/F-actin ratio and decreased G-actin incorporation into junctional actin networks, but it did not change the overall actin expression level or cellular F-actin content. A pharmacological treatment that increased the G/F-actin ratio and decreased actin polymerization mimicked the effects of Tmsb4x depletion on both AJs and PCP. Our results provide insights into the regulation of the actin pool and its involvement in AJ function and PCP establishment.
Sigma-Aldrich
Anti-α-E-Catenin antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-Thymosin beta-4 Antibody, from rabbit, purified by affinity chromatography