Merck
  • The Effect of Low-Temperature Crystallization of Fish Oil on the Chemical Composition, Fatty Acid Profile, and Functional Properties of Cow's Milk.
The Effect of Low-Temperature Crystallization of Fish Oil on the Chemical Composition, Fatty Acid Profile, and Functional Properties of Cow's Milk.
Animals : an open access journal from MDPI (2020-10-15)
Robert Bodkowski, Katarzyna Czyż, Ewa Sokoła-Wysoczańska, Marzena Janczak, Paulina Cholewińska, Anna Wyrostek
ABSTRACT
The study aimed to investigate the effect of supplementation of fish oil after the process of low-temperature crystallization (LTC-FO) enriched with long-chain polyunsaturated fatty acids (LC-PUFAs) on cow milk parameters. The experiment was carried out on 24 Polish Holstein Friesian cows. For 4 weeks, experimental (EXP) group animals (n = 12) were fed LTC-FO (1% of dry matter). Milk was collected two times: on days 14 and 30. LTC-FO supplementation decreased milk fat yield and concentration (p < 0.01). Higher levels of polyunsaturated fatty acids (PUFAs), including these with beneficial biological properties, i.e., eicosapentaenoic (EPA), docosahexaenoic (DHA), docosapentaenoic (DPA), CLA, alpha-linolenic acid (ALA), and TVA (p < 0.01), and lower levels of SFAs, especially short- (p < 0.01) and medium-chain ones (p < 0.05, p < 0.01), were found in the EXP group. The addition of LTC-FO reduced the value of atherogenic and thrombogenic indices as well as SFA/UFA and n-6/n-3 ratios and increased the content of n-3 PUFA and functional fatty acids (p < 0.01). The addition of LTC-FO also increased the delta-9 desaturase index for CLA/TVA and decreased it for pairs C14:1/C14:0 and C16:1/C16:0 (p < 0.05, p < 0.01).
Sigma-Aldrich
cis-4,7,10,13,16,19-Docosahexaenoic acid methyl ester, ≥98%
Sigma-Aldrich
Linoleic acid, conjugated methyl ester
Sigma-Aldrich
Methyl heptadecanoate, ≥99%
Supelco
Methyl all-cis-7,10,13,16,19-docosapentaenoate, analytical standard
Sigma-Aldrich
Methyl all-cis-7,10,13,16,19-docosapentaenoate, ≥97%, liquid