Merck
  • Design of intrinsically stretchable and highly conductive polymers for fully stretchable electrochromic devices.
Design of intrinsically stretchable and highly conductive polymers for fully stretchable electrochromic devices.
Scientific reports (2020-10-07)
Youngno Kim, Chanil Park, Soeun Im, Jung Hyun Kim
ABSTRACT
Stretchable materials are essential for next generation wearable and stretchable electronic devices. Intrinsically stretchable and highly conductive polymers (termed ISHCP) are designed with semi interpenetrating polymer networks (semi-IPN) that enable polymers to be simultaneously applied to transparent electrodes and electrochromic materials. Through a facile method of acid-catalyzed polymer condensation reaction, optimized ISHCP films show the highest electrical conductivity, 1406 S/cm, at a 20% stretched state. Without the blending of any other elastomeric matrix, ISHCP maintains its initial electrical properties under a cyclic stretch-release of over 50% strain. A fully stretchable electrochromic device based on ISHCP is fabricated and shows a performance of 47.7% ∆T and high coloration efficiency of 434.1 cm2/C at 590 nm. The device remains at 45.2% ∆T after 50% strain stretching. A simple patterned electrolyte layer on a stretchable electrochromic device is also realized. The fabricated device, consisting of all-plastic, can be applied by a solution process for large scale production. The ISHCP reveals its potential application in stretchable electrochromic devices and satisfies the requirements for next-generation stretchable electronics.
Sigma-Aldrich
1-Hydroxycyclohexyl phenyl ketone, 99%
Sigma-Aldrich
3,4-Ethylenedioxythiophene, 97%
Sigma-Aldrich
N,N,N′,N′-Tetramethyl-p-phenylenediamine, 99%, powder