Merck
  • Ca2+ plays an antiviral role by increasing p53 expression to achieve protection against spring viraemia of carp virus infection.
Ca2+ plays an antiviral role by increasing p53 expression to achieve protection against spring viraemia of carp virus infection.
Fish & shellfish immunology (2020-05-15)
Long-Feng Lu, Xiao-Yu Zhou, Can Zhang, Zhuo-Cong Li, Dan-Dan Chen, Yong-An Zhang, Shun Li
ABSTRACT
Calcium (Ca) is a messenger that regulates a multitude of physiological processes, but its functions in antiviral progress remain undefined. In this study, we found that Ca2+ enhances fish survival to defend against spring viraemia of carp virus (SVCV) infection by reversing the instability of p53 mediated by the viral protein. First, Ca2+ significantly protected cells and fish against SVCV infection by inducing early apoptosis. Additionally, p53 expression, which was inhibited by SVCV N protein, was upregulated by Ca2+ treatment. Then, the mechanism underlying the reduction of K63-linked p53 ubiquitination by SVCV N protein via the K358 site was completely prevented by Ca2+. These findings reveal the role of Ca2+ in lower vertebrates in the antiviral response, which is connected to and corresponds with viral immune evasion, providing a solution to fish diseases caused by pathogens.
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder
Sigma-Aldrich
Copper(II) chloride dihydrate, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Manganese(II) chloride tetrahydrate, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%
Sigma-Aldrich
Calcium L-lactate hydrate, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
MG-132, Ready Made Solution, ≥90% (HPLC)
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)