Merck
  • Variation among intact tissue samples reveals the core transcriptional features of human CNS cell classes.
Variation among intact tissue samples reveals the core transcriptional features of human CNS cell classes.
Nature neuroscience (2018-08-30)
Kevin W Kelley, Hiromi Nakao-Inoue, Anna V Molofsky, Michael C Oldham
ABSTRACT
It is widely assumed that cells must be physically isolated to study their molecular profiles. However, intact tissue samples naturally exhibit variation in cellular composition, which drives covariation of cell-class-specific molecular features. By analyzing transcriptional covariation in 7,221 intact CNS samples from 840 neurotypical individuals, representing billions of cells, we reveal the core transcriptional identities of major CNS cell classes in humans. By modeling intact CNS transcriptomes as a function of variation in cellular composition, we identify cell-class-specific transcriptional differences in Alzheimer's disease, among brain regions, and between species. Among these, we show that PMP2 is expressed by human but not mouse astrocytes and significantly increases mouse astrocyte size upon ectopic expression in vivo, causing them to more closely resemble their human counterparts. Our work is available as an online resource ( http://oldhamlab.ctec.ucsf.edu/ ) and provides a generalizable strategy for determining the core molecular features of cellular identity in intact biological systems.
Sigma-Aldrich
Anti-APBB1IP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-CELF2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-DBNDD2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution